Search
Search

STK40| Gene Symbol: STK40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737362.2 | XP_014592848.1 | 1018 | 92 | ||
| XM_014737363.2 | XP_014592849.1 | 489 | 92 | |||
| XM_023633369.1 | XP_023489137.1 | 837 | 92 | |||
Set Membership: |
|