Search
Search

KIAA0319L| Gene Symbol: KIAA0319L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503679.6 | XP_001503729.1 | 1419 | 102 | ||
| XM_005607204.3 | XP_005607261.1 | 1496 | 102 | |||
| XM_005607206.3 | XP_005607263.1 | 1330 | 102 | |||
| XM_005607207.3 | XP_005607264.1 | 1253 | 102 | |||
| XM_014737290.2 | XP_014592776.1 | 1325 | 102 | |||
| XM_023633557.1 | XP_023489325.1 | 1232 | 102 | |||
| XM_023633562.1 | XP_023489330.1 | 1093 | 102 | |||
Set Membership: |
|