Search
Search

ZNF362| Gene Symbol: ZNF362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023634034.1 | XP_023489802.1 | 700 | 89 | ||
| XM_023634040.1 | XP_023489808.1 | 571 | 89 | |||
| XM_023634043.1 | XP_023489811.1 | 436 | 89 | |||
Set Membership: |
|