Search
Search
| Gene Symbol: LCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023634361.1 | XP_023490129.1 | 597 | 56 | ||
| XM_023634364.1 | XP_023490132.1 | 432 | 56 | |||
| XM_023634370.1 | XP_023490138.1 | 299 | 56 | |||
Set Membership: |
|