Search
Search

PHACTR4| Gene Symbol: PHACTR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607276.3 | XP_005607333.1 | 2618 | 98 | ||
| XM_005607277.3 | XP_005607334.1 | 2612 | 98 | |||
| XM_005607282.3 | XP_005607339.1 | 2066 | 98 | |||
| XM_005607283.3 | XP_005607340.1 | 2060 | 98 | |||
| XM_023635007.1 | XP_023490775.1 | 1909 | 98 | |||
| XM_023635024.1 | XP_023490792.1 | 2084 | 98 | |||
Set Membership: |
|