Search
Search

CEP85| Gene Symbol: CEP85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504093.4 | XP_001504143.1 | 1941 | 91 | ||
| XM_005607317.3 | XP_005607374.1 | 1776 | 91 | |||
| XM_023635420.1 | XP_023491188.1 | 1804 | 91 | |||
| XM_023635421.1 | XP_023491189.1 | 134 | 91 | |||
Set Membership: |
|