Search
Search
| Gene Symbol: LZIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493871.6 | XP_001493921.2 | 301 | 122 | ||
| XM_005607544.3 | XP_005607601.1 | 301 | 122 | |||
| XM_014737652.2 | XP_014593138.1 | 517 | 122 | |||
| XM_023635828.1 | XP_023491596.1 | 301 | 122 | |||
Set Membership: |
|