Search
Search
| Gene Symbol: RERE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737639.2 | XP_014593125.2 | 3176 | 71 | ||
| XM_023635850.1 | XP_023491618.1 | 2191 | 71 | |||
| XM_023635851.1 | XP_023491619.1 | 2055 | 71 | |||
| XM_023635852.1 | XP_023491620.1 | 3185 | 71 | |||
| XM_023635853.1 | XP_023491621.1 | 3050 | 71 | |||
| XM_023635854.1 | XP_023491622.1 | 1757 | 71 | |||
| XM_023635855.1 | XP_023491623.1 | 725 | 71 | |||
| XM_023635856.1 | XP_023491624.1 | 539 | 71 | |||
| XM_023635857.1 | XP_023491625.1 | 419 | 71 | |||
Set Membership: |
|