Search
Search

DNAJC11| Gene Symbol: DNAJC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023635866.1 | XP_023491634.1 | 1445 | 63 | ||
| XM_023635867.1 | XP_023491635.1 | 1445 | 63 | |||
| XM_023635868.1 | XP_023491636.1 | 1584 | 63 | |||
Set Membership: |
|