Search
Search

ADAMDEC1| Gene Symbol: ADAMDEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607688.3 | XP_005607745.1 | 338 | 118 | ||
| XM_014737821.2 | XP_014593307.1 | 337 | 118 | |||
| XM_023636174.1 | XP_023491942.1 | 336 | 118 | |||
Set Membership: |
|