Search
Search

C2H4orf33| Gene Symbol: C2H4orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607850.3 | XP_005607907.1 | 1558 | 115 | ||
| XM_023636553.1 | XP_023492321.1 | 1479 | 115 | |||
| XM_023636554.1 | XP_023492322.1 | 428 | 115 | |||
Set Membership: |
|