Search
Search
| Gene Symbol: PPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636786.1 | XP_023492554.1 | 1250 | 78 | ||
| XM_023636787.1 | XP_023492555.1 | 1246 | 78 | |||
| XM_023636789.1 | XP_023492557.1 | 560 | 78 | |||
| XM_023636790.1 | XP_023492558.1 | 713 | 78 | |||
| GenBank mRNA | JL637356.1 | - | 4 - 5 | 320 | 78 | |
Set Membership: |
|