Search
Search

TFAP2A| Gene Symbol: TFAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001491087.6 | XP_001491137.1 | 933 | 63 | ||
| XM_003363693.4 | XP_003363741.1 | 1713 | 63 | |||
| XM_023624437.1 | XP_023480205.1 | 1141 | 63 | |||
| XM_023624439.1 | XP_023480207.1 | 1043 | 63 | |||
| XM_023624440.1 | XP_023480208.1 | 1141 | 63 | |||
Set Membership: |
|