Search
Search
| Gene Symbol: GMNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495614.4 | XP_001495664.2 | 330 | 83 | ||
| XM_023624531.1 | XP_023480299.1 | 156 | 83 | |||
| XM_023624532.1 | XP_023480300.1 | 324 | 83 | |||