Search
Search

GABBR1| Gene Symbol: GABBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624578.1 | XP_023480346.1 | 2454 | 82 | ||
| XM_023624579.1 | XP_023480347.1 | 2110 | 82 | |||
| XM_023624580.1 | XP_023480348.1 | 1978 | 82 | |||
| XM_023624581.1 | XP_023480349.1 | 1924 | 82 | |||
Set Membership: |
|