Search
Search

PPP1R10| Gene Symbol: PPP1R10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624174.1 | XP_023479942.1 | 2720 | 115 | ||
| XM_023624175.1 | XP_023479943.1 | 2716 | 115 | |||
| XM_023624176.1 | XP_023479944.1 | 2650 | 115 | |||
| XM_023624177.1 | XP_023479945.1 | 2663 | 115 | |||
| XM_023624178.1 | XP_023479946.1 | 4885 | 115 | |||
| XM_023624179.1 | XP_023479947.1 | 2703 | 115 | |||
Set Membership: |
|