Search
Search
| Gene Symbol: MDFI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496649.6 | XP_001496699.3 | 366 | 94 | ||
| XM_005603900.3 | XP_005603957.1 | 303 | 94 | |||
| XM_005603901.3 | XP_005603958.1 | 166 | 94 | |||