Search
Search

SLC29A1| Gene Symbol: SLC29A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603960.3 | XP_005604017.1 | 287 | 84 | ||
| XM_005603962.3 | XP_005604019.1 | 273 | 84 | |||
| XM_005603963.3 | XP_005604020.1 | 303 | 84 | |||
| XM_023625016.1 | XP_023480784.1 | 322 | 84 | |||
| XM_023625017.1 | XP_023480785.1 | 335 | 84 | |||
| XM_023625018.1 | XP_023480786.1 | 299 | 84 | |||
| XM_023625019.1 | XP_023480787.1 | 284 | 84 | |||
Set Membership: |
|