Search
Search
| Gene Symbol: PHF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625128.1 | XP_023480896.1 | 2871 | 105 | ||
| XM_023625129.1 | XP_023480897.1 | 2738 | 105 | |||
| XM_023625130.1 | XP_023480898.1 | 2862 | 105 | |||
| XM_023625131.1 | XP_023480899.1 | 2805 | 105 | |||
| XM_023625132.1 | XP_023480900.1 | 2744 | 105 | |||
| XM_023625133.1 | XP_023480901.1 | 2583 | 105 | |||
Set Membership: |
|