Search
Search

SIN3B| Gene Symbol: SIN3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734762.2 | XP_014590248.1 | 1006 | 61 | ||
| XM_014734764.2 | XP_014590250.1 | 564 | 61 | |||
| XM_023625378.1 | XP_023481146.1 | 867 | 61 | |||
| XM_023625379.1 | XP_023481147.1 | 999 | 61 | |||
Set Membership: |
|