Search
Search

HOMER3| Gene Symbol: HOMER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625527.1 | XP_023481295.1 | 756 | 74 | ||
| XM_023625528.1 | XP_023481296.1 | 827 | 74 | |||
| XM_023625529.1 | XP_023481297.1 | 854 | 74 | |||
Set Membership: |
|