Search
Search
| Gene Symbol: DDX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604260.3 | XP_005604317.1 | 1586 | 102 | ||
| XM_005604263.3 | XP_005604320.1 | 1515 | 102 | |||
| XM_005604264.3 | XP_005604321.1 | 1484 | 102 | |||
| XM_014734871.2 | XP_014590357.1 | 1413 | 102 | |||
| XM_023625651.1 | XP_023481419.1 | 1506 | 102 | |||
| XM_023625652.1 | XP_023481420.1 | 1587 | 102 | |||
| XM_023625653.1 | XP_023481421.1 | 1542 | 102 | |||
| XM_023625654.1 | XP_023481422.1 | 1527 | 102 | |||
| XM_023625655.1 | XP_023481423.1 | 1509 | 102 | |||
| XM_023625656.1 | XP_023481424.1 | 1510 | 102 | |||
| XM_023625657.1 | XP_023481425.1 | 1482 | 102 | |||
| XM_023625658.1 | XP_023481426.1 | 1455 | 102 | |||
| XM_023625659.1 | XP_023481427.1 | 1449 | 102 | |||
| XM_023625660.1 | XP_023481428.1 | 1438 | 102 | |||
| XM_023625661.1 | XP_023481429.1 | 1410 | 102 | |||
| XM_023625662.1 | XP_023481430.1 | 1336 | 102 | |||