Search
Search

PARP8| Gene Symbol: PARP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604281.3 | XP_005604338.1 | 1129 | 96 | ||
| XM_005604282.3 | XP_005604339.1 | 1130 | 96 | |||
| XM_005604283.3 | XP_005604340.1 | 1130 | 96 | |||
| XM_014734884.2 | XP_014590370.1 | 1040 | 96 | |||