Search
Search

CEP72| Gene Symbol: CEP72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604371.3 | XP_005604428.1 | 2687 | 91 | ||
| XM_005604372.3 | XP_005604429.1 | 2125 | 91 | |||
| XM_023625899.1 | XP_023481667.1 | 1699 | 91 | |||