Search
Search

ENTPD6| Gene Symbol: ENTPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001487835.6 | XP_001487885.3 | 1079 | 74 | ||
| XM_005604418.3 | XP_005604475.1 | 876 | 74 | |||
| XM_023625954.1 | XP_023481722.1 | 876 | 74 | |||
| XM_023625955.1 | XP_023481723.1 | 2642 | 74 | |||
| XM_023625956.1 | XP_023481724.1 | 847 | 74 | |||
Set Membership: |
|