Search
Search
| Gene Symbol: PYGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625957.1 | XP_023481725.1 | 1764 | 99 | ||
| XM_023625958.1 | XP_023481726.1 | 1662 | 99 | |||
| XM_023625959.1 | XP_023481727.1 | 1650 | 99 | |||
Set Membership: |
|