Search
Search
| Gene Symbol: FAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014740697.2 | XP_014596183.1 | 1010 | 95 | ||
| XM_014740698.2 | XP_014596184.1 | 1122 | 95 | |||
| XM_014740700.2 | XP_014596186.1 | 1323 | 95 | |||
| XM_023643306.1 | XP_023499074.1 | 1322 | 95 | |||