Search
Search

PDZRN4| Gene Symbol: PDZRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001914682.5 | XP_001914717.1 | 1429 | 75 | ||
| XM_023643376.1 | XP_023499144.1 | 1397 | 75 | |||
| XM_023643377.1 | XP_023499145.1 | 1310 | 75 | |||