Search
Search

PUS7L| Gene Symbol: PUS7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489281.6 | XP_001489331.3 | 1017 | 77 | ||
| XM_014740935.2 | XP_014596421.1 | 1037 | 77 | |||
| XM_023643401.1 | XP_023499169.1 | 1001 | 77 | |||
| XM_023643402.1 | XP_023499170.1 | 1001 | 77 | |||
| XM_023643403.1 | XP_023499171.1 | 1001 | 77 | |||
| GenBank mRNA | JL638155.1 | - | 2 - 3 | 977 | 77 | |
Set Membership: |
|