Search
Search

ARID2| Gene Symbol: ARID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643424.1 | XP_023499192.1 | 5085 | 86 | ||
| XM_023643426.1 | XP_023499194.1 | 4971 | 86 | |||
| XM_023643427.1 | XP_023499195.1 | 4957 | 86 | |||