Search
Search
| Gene Symbol: ITCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916721.5 | XP_001916756.1 | 894 | 66 | ||
| XM_005604580.3 | XP_005604637.1 | 1020 | 66 | |||
| XM_005604581.3 | XP_005604638.1 | 881 | 66 | |||
| XM_005604582.3 | XP_005604639.1 | 819 | 66 | |||
Set Membership: |
|