Search
Search

NCOA6| Gene Symbol: NCOA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626395.1 | XP_023482163.1 | 6325 | 89 | ||
| XM_023626396.1 | XP_023482164.1 | 6330 | 89 | |||
| XM_023626397.1 | XP_023482165.1 | 6401 | 89 | |||
| XM_023626398.1 | XP_023482166.1 | 6251 | 89 | |||
| XM_023626399.1 | XP_023482167.1 | 6256 | 89 | |||
| XM_023626400.1 | XP_023482168.1 | 6325 | 89 | |||
| XM_023626401.1 | XP_023482169.1 | 6292 | 89 | |||
| XM_023626402.1 | XP_023482170.1 | 6289 | 89 | |||
| XM_023626403.1 | XP_023482171.1 | 6025 | 89 | |||
| GenBank mRNA | JL620767.1 | - | 1 - 2 | 66 | 89 | |
Set Membership: |
|