Search
Search

EPB41L1| Gene Symbol: EPB41L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626478.1 | XP_023482246.1 | 585 | 116 | ||
| XM_023626479.1 | XP_023482247.1 | 585 | 116 | |||
| XM_023626480.1 | XP_023482248.1 | 585 | 116 | |||
| XM_023626481.1 | XP_023482249.1 | 585 | 116 | |||
| XM_023626482.1 | XP_023482250.1 | 585 | 116 | |||
| XM_023626483.1 | XP_023482251.1 | 722 | 116 | |||
| XM_023626485.1 | XP_023482253.1 | 778 | 116 | |||
| XM_023626486.1 | XP_023482254.1 | 934 | 116 | |||
| XM_023626487.1 | XP_023482255.1 | 869 | 116 | |||
| XM_023626488.1 | XP_023482256.1 | 849 | 116 | |||
| XM_023626489.1 | XP_023482257.1 | 656 | 116 | |||
| XM_023626490.1 | XP_023482258.1 | 940 | 116 | |||
| XM_023626491.1 | XP_023482259.1 | 724 | 116 | |||
| XM_023626492.1 | XP_023482260.1 | 786 | 116 | |||
| XM_023626493.1 | XP_023482261.1 | 585 | 116 | |||
| XM_023626494.1 | XP_023482262.1 | 585 | 116 | |||
| XM_023626495.1 | XP_023482263.1 | 585 | 116 | |||
| XM_023626496.1 | XP_023482264.1 | 585 | 116 | |||
| XM_023626498.1 | XP_023482266.1 | 585 | 116 | |||
| XM_023626499.1 | XP_023482267.1 | 585 | 116 | |||
Set Membership: |
|