Search
Search

PLCG1| Gene Symbol: PLCG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604632.3 | XP_005604689.1 | 304 | 74 | ||
| XM_023626566.1 | XP_023482334.1 | 537 | 74 | |||
| XM_023626567.1 | XP_023482335.1 | 609 | 74 | |||
Set Membership: |
|