Search
Search

ZNF335| Gene Symbol: ZNF335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503381.4 | XP_001503431.1 | 3636 | 82 | ||
| XM_014735186.2 | XP_014590672.1 | 3979 | 82 | |||
| XM_014735187.2 | XP_014590673.1 | 3975 | 82 | |||
Set Membership: |
|