Search
Search

CSE1L| Gene Symbol: CSE1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503546.6 | XP_001503596.2 | 2583 | 137 | ||
| XM_023626699.1 | XP_023482467.1 | 2561 | 137 | |||
| XM_023626700.1 | XP_023482468.1 | 2472 | 137 | |||
Set Membership: |
|