Search
Search

DIDO1| Gene Symbol: DIDO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001491593.4 | XP_001491643.2 | 1453 | 83 | ||
| XM_005604850.3 | XP_005604907.1 | 1558 | 83 | |||
| XM_005604851.3 | XP_005604908.1 | 1537 | 83 | |||
| XM_014735325.2 | XP_014590811.1 | 1549 | 83 | |||
| XM_023626891.1 | XP_023482659.1 | 1448 | 83 | |||
| XM_023626892.1 | XP_023482660.1 | 1578 | 83 | |||
Set Membership: |
|