Search
Search

SLC17A9| Gene Symbol: SLC17A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604852.3 | XP_005604909.1 | 1051 | 67 | ||
| XM_023626898.1 | XP_023482666.1 | 1052 | 67 | |||
| XM_023626902.1 | XP_023482670.1 | 949 | 67 | |||
Set Membership: |
|