Search
Search

TPD52L2| Gene Symbol: TPD52L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001915620.4 | XP_001915655.1 | 303 | 86 | ||
| XM_003363952.4 | XP_003364000.1 | 303 | 86 | |||
| XM_005604862.3 | XP_005604919.1 | 304 | 86 | |||
| XM_005604863.3 | XP_005604920.1 | 303 | 86 | |||
| XM_005604864.3 | XP_005604921.1 | 304 | 86 | |||
| XM_005604865.3 | XP_005604922.1 | 304 | 86 | |||
Set Membership: |
|