Search
Search

NAA35| Gene Symbol: NAA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023627118.1 | XP_023482886.1 | 1715 | 103 | ||
| XM_023627119.1 | XP_023482887.1 | 1710 | 103 | |||
| XM_023627120.1 | XP_023482888.1 | 1653 | 103 | |||
| XM_023627121.1 | XP_023482889.1 | 1678 | 103 | |||
Set Membership: |
|