Search
Search
| Gene Symbol: IL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604993.3 | XP_005605050.1 | 159 | 98 | ||
| XM_014735451.2 | XP_014590937.1 | 149 | 98 | |||
| XM_023627334.1 | XP_023483102.1 | 173 | 98 | |||
Set Membership: |
|