Search
Search
| Gene Symbol: FGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605105.3 | XP_005605162.1 | 962 | 107 | ||
| XM_014735538.2 | XP_014591024.1 | 1162 | 107 | |||
| XM_023627614.1 | XP_023483382.1 | 1055 | 107 | |||
| XM_023627615.1 | XP_023483383.1 | 1971 | 107 | |||
| XM_023627616.1 | XP_023483384.1 | 883 | 107 | |||
Set Membership: |
|