Search
Search

PLEKHG3| Gene Symbol: PLEKHG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014735639.2 | XP_014591125.2 | 1248 | 70 | ||
| XM_023627912.1 | XP_023483680.1 | 1249 | 70 | |||
| XM_023627913.1 | XP_023483681.1 | 1249 | 70 | |||
| XM_023627914.1 | XP_023483682.1 | 1364 | 70 | |||
| XM_023627915.1 | XP_023483683.1 | 1168 | 70 | |||
| XM_023627916.1 | XP_023483684.1 | 1243 | 70 | |||
Set Membership: |
|