Search
Search

EIF2S1| Gene Symbol: EIF2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499768.4 | XP_001499818.1 | 521 | 83 | ||
| XM_023627953.1 | XP_023483721.1 | 401 | 83 | |||
| GenBank mRNA | JL616195.1 | - | 2 - 3 | 368 | 83 | |
Set Membership: |
|