Search
Search

ZFYVE26| Gene Symbol: ZFYVE26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500061.5 | XP_001500111.3 | 1428 | 81 | ||
| XM_014735666.2 | XP_014591152.2 | 1267 | 81 | |||
| XM_023627976.1 | XP_023483744.1 | 1428 | 81 | |||