Search
Search

GPATCH2L| Gene Symbol: GPATCH2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628147.1 | XP_023483915.1 | 775 | 65 | ||
| XM_023628148.1 | XP_023483916.1 | 775 | 65 | |||
| XM_023628149.1 | XP_023483917.1 | 775 | 65 | |||
Set Membership: |
|