Search
Search

ADCK1| Gene Symbol: ADCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494170.6 | XP_001494220.4 | 727 | 69 | ||
| XM_023628190.1 | XP_023483958.1 | 891 | 69 | |||
| XM_023628191.1 | XP_023483959.1 | 898 | 69 | |||
| XM_023628192.1 | XP_023483960.1 | 1851 | 69 | |||
| XM_023628194.1 | XP_023483962.1 | 523 | 69 | |||
Set Membership: |
|