Search
Search
| Gene Symbol: VRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628356.1 | XP_023484124.1 | 1029 | 89 | ||
| XM_023628357.1 | XP_023484125.1 | 1029 | 89 | |||
| XM_023628358.1 | XP_023484126.1 | 1029 | 89 | |||
| XM_023628359.1 | XP_023484127.1 | 1029 | 89 | |||
| GenBank mRNA | HQ890083.1 | - | 5 - 6 | 480 | 89 | |
| JL616215.2 | - | 10 - 11 | 976 | 89 | ||
Set Membership: |
|