Search
Search

WARS1| Gene Symbol: WARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488721.6 | XP_001488771.3 | 1496 | 89 | ||
| XM_023628399.1 | XP_023484167.1 | 1385 | 89 | |||
| XM_023628400.1 | XP_023484168.1 | 1418 | 89 | |||
Set Membership: |
|