Search
Search

CDC42BPB| Gene Symbol: CDC42BPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628441.1 | XP_023484209.1 | 5188 | 60 | ||
| XM_023628442.1 | XP_023484210.1 | 5118 | 60 | |||
| XM_023628443.1 | XP_023484211.1 | 5078 | 60 | |||
Set Membership: |
|